Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZZZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015534.5 | NP_056349.1 | 5 | 1141 | ||
XM_005270725.3 | XP_005270782.1 | 5 | 1216 | |||
XM_005270726.3 | XP_005270783.1 | 4 | 1117 | |||
XM_005270727.3 | XP_005270784.1 | 5 | 1216 | |||
XM_017000970.1 | XP_016856459.1 | 4 | 1117 | |||
XM_017000971.1 | XP_016856460.1 | 5 | 1216 | |||
GenBank mRNA | AK074119.1 | - | 1 | 187 | ||
AK310514.1 | - | 5 | 1138 | |||
BC035818.2 | - | 1 | 716 | 5554765 | ||
BX640766.1 | - | 1 | 418 | |||
BX641001.1 | - | 5 | 1142 | |||
BX641096.1 | - | 5 | 1127 |