Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATS2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100422.1 | NP_001093892.1 | 8 | 1115 | ||
NM_001100423.1 | NP_001093893.1 | 8 | 1065 | |||
NM_001100424.1 | NP_001093894.1 | 7 | 908 | |||
NM_001282735.1 | NP_001269664.1 | 8 | 1187 | |||
NM_001282743.1 | NP_001269672.1 | 7 | 1126 | |||
NM_001282744.1 | NP_001269673.1 | 8 | 916 | |||
NM_015535.2 | NP_056350.2 | 8 | 1055 | |||
XM_005246456.1 | XP_005246513.1 | 9 | 974 | |||
XM_005246457.1 | XP_005246514.1 | 9 | 1237 | |||
XM_005246458.1 | XP_005246515.1 | 7 | 848 | |||
XM_005246459.1 | XP_005246516.1 | 7 | 655 | |||
XM_011510938.1 | XP_011509240.1 | 9 | 1205 | |||
XM_011510939.1 | XP_011509241.1 | 8 | 918 | |||
XM_017003783.1 | XP_016859272.1 | 9 | 1069 | |||
XM_017003784.1 | XP_016859273.1 | 7 | 3183 | |||
XM_017003785.1 | XP_016859274.1 | 6 | 641 | |||
XM_017003786.1 | XP_016859275.1 | 8 | 767 | |||
XM_017003787.1 | XP_016859276.1 | 8 | 1030 | |||
GenBank mRNA | AF193059.1 | - | 8 | 1001 | ||
AK002064.1 | - | 8 | 853 | |||
AK090867.1 | - | 7 | 848 | |||
AK290496.1 | - | 8 | 1175 | |||
AK299223.1 | - | 7 | 828 | |||
AK300075.1 | - | 8 | 918 | |||
AK314561.1 | - | 7 | 616 | |||
AL110124.1 | - | 8 | 863 | |||
AL525852.3 | - | 7 | 835 | |||
AY450394.1 | - | 8 | 863 | |||
BC018736.1 | - | 7 | 601 | 4845423 |