Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM32A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014077.3 | NP_054796.1 | 3 | 286 | ||
GenBank mRNA | AF151902.1 | - | 3 | 245 | ||
AK312058.1 | - | 3 | 269 | |||
AL050157.1 | - | 3 | 264 | |||
BC000639.1 | - | 3 | 264 | 3345441 | ||
BC004447.1 | - | 2 | 116 | |||
BC017286.1 | - | 3 | 247 | 4892079 | ||
BG749750.1 | - | 2 | 413 |