Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015542.3 | NP_056357.1 | 14 | 2917 | ||
NM_080599.2 | NP_542166.1 | 14 | 3087 | |||
XM_017016034.1 | XP_016871523.1 | 13 | 2573 | |||
GenBank mRNA | AB037829.1 | - | 13 | 3286 | ||
AF301013.1 | - | 14 | 2910 | |||
AF318574.1 | - | 14 | 2887 | |||
AK310925.1 | - | 14 | 2876 | |||
AL080198.1 | - | 8 | 1087 | |||
AY013249.1 | - | 13 | 2836 | |||
BC114964.1 | - | 14 | 2881 | 40082739 | ||
BC115737.1 | - | 14 | 2881 | 40082728 |