Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015542.3 | NP_056357.1 | 5 | 1503 | ||
NM_080599.2 | NP_542166.1 | 5 | 1673 | |||
XM_011519449.2 | XP_011517751.1 | 5 | 1551 | |||
XM_017016034.1 | XP_016871523.1 | 4 | 1159 | |||
GenBank mRNA | AB037829.1 | - | 4 | 1872 | ||
AF301013.1 | - | 5 | 1496 | |||
AF318574.1 | - | 5 | 1473 | |||
AK096191.1 | - | 6 | 1413 | |||
AK310925.1 | - | 5 | 1462 | |||
AY013249.1 | - | 4 | 1422 | |||
BC114964.1 | - | 5 | 1467 | 40082739 | ||
BC115737.1 | - | 5 | 1467 | 40082728 |