Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015550.3 | NP_056365.1 | 21 | 2816 | ||
NM_145320.2 | NP_663160.1 | 19 | 2396 | |||
NM_145321.2 | NP_663161.1 | 19 | 2381 | |||
NM_145322.2 | NP_663162.1 | 18 | 2288 | |||
NR_104111.1 | 20 | 2888 | ||||
NR_104112.1 | 19 | 2795 | ||||
XM_005249698.3 | XP_005249755.1 | 21 | 2536 | |||
XM_006715681.3 | XP_006715744.1 | 20 | 2710 | |||
XM_006715682.3 | XP_006715745.1 | 20 | 2714 | |||
XM_006715683.3 | XP_006715746.1 | 19 | 2621 | |||
XM_006715684.3 | XP_006715747.1 | 20 | 2473 | |||
XM_011515258.2 | XP_011513560.1 | 21 | 2613 | |||
XM_011515259.2 | XP_011513561.1 | 22 | 3110 | |||
XM_017011948.1 | XP_016867437.1 | 23 | 2973 | |||
XM_017011949.1 | XP_016867438.1 | 21 | 3002 | |||
GenBank mRNA | AB014604.2 | - | 21 | 2753 | ||
AF392444.1 | - | 21 | 2725 | |||
AF491781.1 | - | 20 | 2443 | |||
AF491782.1 | - | 19 | 2350 | |||
AF491783.1 | - | 19 | 2335 | |||
AF491784.1 | - | 18 | 2242 | |||
AF491785.1 | - | 20 | 2842 | |||
AF491786.1 | - | 19 | 2749 | |||
AL832293.1 | - | 18 | 2052 | |||
AY008372.1 | - | 21 | 2646 | |||
BC017731.1 | - | 21 | 2745 | 3909164 |