Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015550.3 | NP_056365.1 | 15 | 1987 | ||
NM_145320.2 | NP_663160.1 | 13 | 1567 | |||
NM_145321.2 | NP_663161.1 | 13 | 1552 | |||
NM_145322.2 | NP_663162.1 | 12 | 1459 | |||
NR_104111.1 | 14 | 1660 | ||||
NR_104112.1 | 13 | 1567 | ||||
XM_005249698.3 | XP_005249755.1 | 15 | 1707 | |||
XM_006715681.3 | XP_006715744.1 | 14 | 1881 | |||
XM_006715682.3 | XP_006715745.1 | 14 | 1885 | |||
XM_006715683.3 | XP_006715746.1 | 13 | 1792 | |||
XM_006715684.3 | XP_006715747.1 | 14 | 1644 | |||
XM_011515258.2 | XP_011513560.1 | 15 | 1784 | |||
XM_011515259.2 | XP_011513561.1 | 16 | 2281 | |||
XM_017011948.1 | XP_016867437.1 | 17 | 2144 | |||
XM_017011949.1 | XP_016867438.1 | 15 | 2173 | |||
GenBank mRNA | AB014604.2 | - | 15 | 1924 | ||
AF392444.1 | - | 15 | 1896 | |||
AF491781.1 | - | 14 | 1614 | |||
AF491782.1 | - | 13 | 1521 | |||
AF491783.1 | - | 13 | 1506 | |||
AF491784.1 | - | 12 | 1413 | |||
AF491785.1 | - | 14 | 1614 | |||
AF491786.1 | - | 13 | 1521 | |||
AF515639.1 | - | 7 | 666 | |||
AF515640.1 | - | 6 | 573 | |||
AL832293.1 | - | 12 | 1223 | |||
AY008372.1 | - | 15 | 1817 | |||
BC017731.1 | - | 15 | 1916 | 3909164 |