Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324091.1 | NP_001311020.1 | 4 | 1070 | ||
NM_001324092.1 | NP_001311021.1 | 3 | 471 | |||
NM_001324093.1 | NP_001311022.1 | 5 | 1197 | |||
NM_001324094.1 | NP_001311023.1 | 5 | 2197 | |||
NM_015554.2 | NP_056369.1 | 4 | 1070 | |||
XM_005254298.3 | XP_005254355.1 | 5 | 1202 | |||
XM_017022073.1 | XP_016877562.1 | 3 | 1054 | |||
XM_017022074.1 | XP_016877563.1 | 3 | 1044 | |||
GenBank mRNA | AB020643.1 | - | 2 | 782 | ||
AK002188.1 | - | 2 | 394 | |||
AY635582.1 | - | 4 | 1033 |