Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324092.1 | NP_001311021.1 | 4 | 906 | ||
NM_001324093.1 | NP_001311022.1 | 6 | 1632 | |||
NM_001324094.1 | NP_001311023.1 | 6 | 2632 | |||
NM_015554.2 | NP_056369.1 | 5 | 1505 | |||
XM_005254298.3 | XP_005254355.1 | 6 | 1637 | |||
XM_017022073.1 | XP_016877562.1 | 4 | 1489 | |||
XM_017022074.1 | XP_016877563.1 | 4 | 2924 | |||
GenBank mRNA | AB020643.1 | - | 3 | 1217 | ||
AK002188.1 | - | 3 | 829 | |||
AY635582.1 | - | 5 | 1468 |