Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015129.5 | NP_055944.2 | 5 | 842 | ||
NM_145799.3 | NP_665798.1 | 5 | 842 | |||
NM_145800.3 | NP_665799.1 | 5 | 842 | |||
NM_145802.3 | NP_665801.1 | 5 | 842 | |||
XM_005262400.3 | XP_005262457.1 | 5 | 650 | |||
XM_006724748.2 | XP_006724811.1 | 5 | 650 | |||
XM_006724750.2 | XP_006724813.1 | 5 | 650 | |||
XM_011531317.2 | XP_011529619.1 | 5 | 650 | |||
XM_011531318.2 | XP_011529620.1 | 5 | 650 | |||
GenBank mRNA | AF397023.1 | - | 5 | 687 | ||
AF403058.1 | - | 5 | 842 | |||
AF403059.1 | - | 5 | 842 | |||
AF403060.1 | - | 5 | 842 | |||
AF403061.1 | - | 5 | 842 | |||
AF403062.1 | - | 5 | 842 | |||
AK026589.1 | - | 5 | 635 | |||
AK290977.1 | - | 5 | 894 | |||
AK298487.1 | - | 6 | 749 | |||
AK299273.1 | - | 5 | 697 | |||
AK303116.1 | - | 6 | 719 | |||
AK303224.1 | - | 5 | 765 | |||
AY005981.1 | - | 5 | 734 | |||
AY034177.1 | - | 5 | 832 | |||
AY679521.1 | - | 5 | 842 | |||
BC009291.2 | - | 5 | 626 | 4111673 | ||
BC011922.2 | - | 5 | 630 | 4542179 | ||
BC036240.1 | - | 5 | 669 | 5298284 | ||
BC069231.1 | - | 5 | 614 | 4852319 | ||
D50918.1 | - | 570 |