Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTNAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014141.5 | NP_054860.1 | 4, 5 | 1075 | ||
XM_017011950.1 | XP_016867439.1 | 4, 5 | 1401 | |||
GenBank mRNA | AB020675.1 | - | 4, 5 | 584 | ||
AF319045.1 | - | 4, 5 | 699 | |||
AK315113.1 | - | 4, 5 | 664 | |||
BC093780.1 | - | 4, 5 | 602 | 7939625 | ||
BC113373.1 | - | 4, 5 | 602 | 8322425 | ||
KJ534811.1 | - | 3, 4 | 448 |