Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303450.1 | NP_001290379.1 | 2 | 366 | ||
NM_021646.3 | NP_067678.1 | 2 | 366 | |||
XM_011522453.2 | XP_011520755.1 | 2 | 367 | |||
XM_011522454.2 | XP_011520756.1 | 1 | 164 | |||
GenBank mRNA | AA236649.1 | - | 2 | 277 | ||
AB011129.1 | - | 1 | 106 | |||
AK291792.1 | - | 2 | 310 | |||
AK297993.1 | - | 2 | 365 | |||
BC121013.1 | - | 1 | 92 | 40117042 | ||
BC121014.1 | - | 1 | 92 | 40117044 | ||
CN314802.1 | - | 2 | 339 | |||
DC335253.1 | - | 2 | 368 |