Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303450.1 | NP_001290379.1 | 3 | 769 | ||
NM_021646.3 | NP_067678.1 | 3 | 769 | |||
XM_005255243.3 | XP_005255300.1 | 2 | 735 | |||
XM_011522453.2 | XP_011520755.1 | 3 | 770 | |||
XM_011522454.2 | XP_011520756.1 | 2 | 524 | |||
XM_017023121.1 | XP_016878610.1 | 2 | 262 | |||
GenBank mRNA | AB011129.1 | - | 2 | 509 | ||
AK291792.1 | - | 3 | 713 | |||
AK297993.1 | - | 3 | 768 | |||
BC121013.1 | - | 2 | 495 | 40117042 | ||
BC121014.1 | - | 2 | 452 | 40117044 | ||
CN314802.1 | - | 3 | 700 |