Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000729.5 | NP_000720.1 | 5 | 1317 | ||
NM_001174138.2 | NP_001167609.1 | 3 | 550 | |||
GenBank mRNA | AK289716.1 | - | 3 | 492 | ||
BC008283.1 | - | 3 | 550 | 3639109 | ||
BC093055.1 | - | 3 | 492 | 30722619 | ||
BC111026.1 | - | 5 | 1292 | 5199880 | ||
BI756101.1 | - | 3 | 478 | |||
CB961869.1 | - | 3 | 519 | |||
CD250282.1 | - | 3 | 652 |