Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303450.1 | NP_001290379.1 | 6 | 1402 | ||
NM_021646.3 | NP_067678.1 | 6 | 1402 | |||
XM_005255243.3 | XP_005255300.1 | 5 | 1368 | |||
XM_011522453.2 | XP_011520755.1 | 6 | 1403 | |||
XM_011522454.2 | XP_011520756.1 | 5 | 1157 | |||
XM_017023121.1 | XP_016878610.1 | 5 | 895 | |||
GenBank mRNA | AB011129.1 | - | 5 | 1142 | ||
AK291792.1 | - | 6 | 1346 | |||
AK297993.1 | - | 6 | 1401 | |||
BC004125.2 | - | 1 | 343 | |||
BC033151.1 | - | 1 | 472 | |||
BC121013.1 | - | 5 | 1128 | 40117042 | ||
BC121014.1 | - | 5 | 1085 | 40117044 |