Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286771.2 | NP_001273700.1 | 29 | 6410 | ||
NM_015574.1 | NP_056389.1 | 29 | 6789 | |||
NM_032217.4 | NP_115593.3 | 29 | 6792 | |||
NM_198889.2 | NP_942592.1 | 28 | 6039 | |||
XM_005265671.3 | XP_005265728.1 | 28 | 6380 | |||
XM_017008011.1 | XP_016863500.1 | 29 | 6364 | |||
XM_017008012.1 | XP_016863501.1 | 28 | 5614 | |||
XM_017008013.1 | XP_016863502.1 | 28 | 5611 | |||
GenBank mRNA | AB014597.2 | - | 29 | 6313 | ||
AK096518.1 | - | 5 | 2136 | |||
AL833287.1 | - | 25 | 5216 | |||
BC019963.2 | - | 1 | 1336 | |||
BX648035.1 | - | 29 | 6363 |