Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIGYF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103146.1 | NP_001096616.1 | 10 | 971 | ||
NM_001103147.1 | NP_001096617.1 | 12 | 1107 | |||
NM_001103148.1 | NP_001096618.1 | 9 | 887 | |||
NM_015575.3 | NP_056390.2 | 12 | 1111 | |||
NR_103492.1 | 9 | 887 | ||||
GenBank mRNA | AB014542.2 | - | 12 | 1066 | ||
AK001739.1 | - | 12 | 1085 | |||
AK299418.1 | - | 11 | 1015 | |||
AK302998.1 | - | 4 | 473 | |||
AY176045.1 | - | 12 | 1085 | |||
BC043402.1 | - | 10 | 906 | |||
BC136251.1 | - | 9 | 842 | 9020238 | ||
BC143352.1 | - | 9 | 842 | |||
BC146775.1 | - | 12 | 1066 | 9007224 | ||
BX537885.1 | - | 10 | 906 | |||
BX538172.1 | - | 9 | 855 | |||
BX538321.1 | - | 10 | 927 | |||
BX647505.1 | - | 9 | 718 |