Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012096.2 | NP_036228.1 | 10 | 962 | ||
XM_011533583.2 | XP_011531885.1 | 11 | 1015 | |||
GenBank mRNA | AB037849.2 | - | 10 | 815 | ||
AF169797.1 | - | 10 | 873 | |||
AF424738.1 | - | 10 | 871 | |||
AK299008.1 | - | 11 | 1004 | |||
AK310716.1 | - | 10 | 910 | |||
BC028599.2 | - | 10 | 962 | 4829430 |