Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251962.1 | NP_001238891.1 | 5 | 1574 | ||
NM_001251963.1 | NP_001238892.1 | 5 | 1501 | |||
NM_015595.3 | NP_056410.3 | 5 | 1501 | |||
XM_011512672.1 | XP_011510974.1 | 5 | 1501 | |||
XM_017006160.1 | XP_016861649.1 | 4 | 243 | |||
GenBank mRNA | AF415175.1 | - | 5 | 1456 | ||
AK022884.1 | - | 2 | 88 | |||
AK123040.1 | - | 5 | 1445 | |||
AK127282.1 | - | 5 | 1574 | |||
AY552599.1 | - | 4 | 1290 | |||
BC078655.1 | - | 5 | 1442 | 4374088 |