Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278185.1 | NP_001265114.1 | 14 | 1806 | ||
NM_001278186.1 | NP_001265115.1 | 12 | 1554 | |||
NM_015601.3 | NP_056416.2 | 14 | 1806 | |||
NM_022079.2 | NP_071362.1 | 14 | 1806 | |||
XM_011539592.2 | XP_011537894.1 | 15 | 1880 | |||
XM_011539593.2 | XP_011537895.1 | 15 | 1880 | |||
XM_011539594.2 | XP_011537896.1 | 16 | 1966 | |||
XM_011539595.2 | XP_011537897.1 | 16 | 2016 | |||
XM_011539596.2 | XP_011537898.1 | 15 | 1894 | |||
GenBank mRNA | AB046813.1 | - | 11 | 1660 | ||
AK027835.1 | - | 5 | 373 | |||
AK292809.1 | - | 14 | 1686 | |||
AY221963.1 | - | 14 | 1743 | |||
AY650032.1 | - | 12 | 1491 | |||
AY650033.1 | - | 14 | 1743 | |||
BC015043.1 | - | 12 | 1457 | |||
BC039600.1 | - | 14 | 1716 | 5212048 | ||
BX537563.1 | - | 13 | 4191 |