Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KRIT1 SIRNA Assays ›
Gene Symbol:
KRIT1
Gene Name:
KRIT1, ankyrin repeat containing
Gene Aliases:
CAM, CCM1
Chromosome Location:
Chr.7: 92198969 - 92246128 on Build GRCh38
UniGene ID:
Hs.531987
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s2509
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KRIT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001013406.1
NP_001013424.1
14
2009
NM_004912.3
NP_004903.2
16
2648
NM_194454.1
NP_919436.1
16
2301
NM_194455.1
NP_919437.1
16
2775
NM_194456.1
NP_919438.1
17
2540
XM_005250660.3
XP_005250717.1
16
2268
XM_005250662.3
XP_005250719.1
16
2247
XM_005250665.3
XP_005250722.1
16
2175
XM_005250666.3
XP_005250723.1
17
2351
XM_005250667.2
XP_005250724.1
15
2059
XM_005250668.3
XP_005250725.1
15
2121
XM_005250669.3
XP_005250726.1
15
2099
XM_006716161.3
XP_006716224.1
16
2399
XM_006716162.3
XP_006716225.1
16
2216
XM_006716163.3
XP_006716226.1
15
2066
XM_011516651.2
XP_011514953.1
17
2382
XM_011516653.2
XP_011514955.1
15
2136
XM_011516654.2
XP_011514956.1
17
2400
XM_011516655.2
XP_011514957.1
15
2054
XM_011516656.2
XP_011514958.1
15
2023
XM_011516657.2
XP_011514959.1
15
1943
XM_011516658.2
XP_011514960.1
15
1955
XM_011516659.2
XP_011514961.1
15
1977
XM_011516660.2
XP_011514962.1
14
1859
XM_011516661.2
XP_011514963.1
14
1937
XM_017012755.1
XP_016868244.1
15
2026
XM_017012756.1
XP_016868245.1
15
1988
XM_017012757.1
XP_016868246.1
15
2020
XM_017012758.1
XP_016868247.1
16
2495
XM_017012759.1
XP_016868248.1
15
2347
GenBank mRNA
AF296765.2
-
16
2545
AF310133.1
-
16
2240
AF388384.1
-
16
2775
AK055305.1
-
14
2009
AK056537.1
-
15
2083
AK226131.1
-
9
1398
AY380057.1
-
16
2240
AY380058.1
-
16
2240
AY380059.1
-
16
2239
AY380060.1
-
16
2238
AY993944.1
-
13
1756
BC094684.1
-
13
1766
6174749
BX648659.1
-
10
2280
U90268.1
-
9
1160
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
negative regulation of endothelial cell proliferation
small GTPase mediated signal transduction
negative regulation of endothelial cell migration
negative regulation of angiogenesis
positive regulation of protein binding
cell redox homeostasis
regulation of catalytic activity
regulation of establishment of cell polarity
negative regulation of endothelial cell apoptotic process
protein binding
phosphatidylinositol-4,5-bisphosphate binding
microtubule binding
GTPase regulator activity
protein complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-69b65fdbbd-pb7tv:80/100.66.78.176:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline