Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WIPI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033518.1 | NP_001028690.1 | 7 | 854 | ||
NM_001033519.1 | NP_001028691.1 | 6 | 800 | |||
NM_001033520.1 | NP_001028692.1 | 4 | 693 | |||
NM_001278299.1 | NP_001265228.1 | 7 | 1028 | |||
NM_015610.3 | NP_056425.1 | 7 | 854 | |||
NM_016003.3 | NP_057087.2 | 6 | 800 | |||
XM_006715685.1 | XP_006715748.1 | 5 | 686 | |||
XM_017011952.1 | XP_016867441.1 | 5 | 686 | |||
GenBank mRNA | AF151808.1 | - | 4 | 692 | ||
AK023041.1 | - | 7 | 814 | |||
AK024279.1 | - | 6 | 779 | |||
AK315468.1 | - | 7 | 974 | |||
AL080155.1 | - | 7 | 855 | |||
AY691425.1 | - | 6 | 584 | |||
AY691426.1 | - | 4 | 624 | |||
BC004116.2 | - | 7 | 797 | 3842487 | ||
BC007596.1 | - | 6 | 767 | 3346710 | ||
BC021012.1 | - | 6 | 719 | |||
BC021068.1 | - | 6 | 719 | |||
BC021200.2 | - | 7 | 795 | 3957981 | ||
CR533530.1 | - | 7 | 638 |