Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TANC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025185.3 | NP_079461.2 | 7, 8 | 958 | ||
XM_005257203.4 | XP_005257260.1 | 8, 9 | 1185 | |||
XM_006721806.3 | XP_006721869.1 | 9, 10 | 1407 | |||
XM_006721807.3 | XP_006721870.1 | 9, 10 | 1407 | |||
XM_006721810.3 | XP_006721873.1 | 9, 10 | 1407 | |||
XM_006721811.3 | XP_006721874.1 | 9, 10 | 1407 | |||
XM_011524597.2 | XP_011522899.1 | 9, 10 | 1407 | |||
XM_011524598.2 | XP_011522900.1 | 9, 10 | 1407 | |||
XM_011524599.1 | XP_011522901.1 | 9, 10 | 1233 | |||
XM_011524600.2 | XP_011522902.1 | 8, 9 | 1296 | |||
XM_011524601.2 | XP_011522903.1 | 9, 10 | 1296 | |||
XM_011524602.2 | XP_011522904.1 | 8, 9 | 1185 | |||
XM_017024429.1 | XP_016879918.1 | 9, 10 | 1233 | |||
XM_017024430.1 | XP_016879919.1 | 5, 6 | 4574 | |||
XM_017024431.1 | XP_016879920.1 | 9, 10 | 1407 | |||
XM_017024432.1 | XP_016879921.1 | 9, 10 | 1407 | |||
GenBank mRNA | AB046856.2 | - | 3, 4 | 559 |