Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015630.3 | NP_056445.3 | 9 | 1377 | ||
XM_011510941.2 | XP_011509243.1 | 9 | 1588 | |||
XM_011510943.2 | XP_011509245.1 | 8 | 1261 | |||
GenBank mRNA | AF286904.1 | - | 9 | 1369 | ||
AK001433.1 | - | 7 | 968 | |||
AK125789.1 | - | 10 | 1446 | |||
BC093818.1 | - | 9 | 1289 | 7939663 | ||
BC093820.1 | - | 9 | 1289 | 7939665 |