Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015630.3 | NP_056445.3 | 9, 10 | 1412 | ||
XM_011510941.2 | XP_011509243.1 | 9, 10 | 1623 | |||
XM_011510943.2 | XP_011509245.1 | 8, 9 | 1296 | |||
GenBank mRNA | AF286904.1 | - | 9, 10 | 1404 | ||
AK001433.1 | - | 7, 8 | 1003 | |||
AK125789.1 | - | 10, 11 | 1481 | |||
BC093818.1 | - | 9, 10 | 1324 | 7939663 | ||
BC093820.1 | - | 9, 10 | 1324 | 7939665 |