Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGFR1OP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171887.1 | NP_001165358.1 | 6 | 934 | ||
NM_015633.2 | NP_056448.1 | 7 | 1048 | |||
GenBank mRNA | AF161472.1 | - | 6 | 800 | ||
AK001534.1 | - | 7 | 1037 | |||
AK094888.1 | - | 2 | 2524 | |||
AL117608.1 | - | 4 | 442 | |||
AY506561.1 | - | 5 | 581 |