Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305077.1 | NP_001292006.1 | 6 | 605 | ||
NM_001305078.1 | NP_001292007.1 | 6 | 604 | |||
NM_001305080.1 | NP_001292009.1 | 6 | 593 | |||
NM_001305081.1 | NP_001292010.1 | 6 | 602 | |||
NM_001305082.1 | NP_001292011.1 | 6 | 753 | |||
NM_001305083.1 | NP_001292012.1 | 6 | 590 | |||
NM_015654.4 | NP_056469.2 | 6 | 608 | |||
NR_130953.1 | 7 | 612 | ||||
XM_005257212.4 | XP_005257269.1 | 5 | 447 | |||
XM_006721813.3 | XP_006721876.1 | 5 | 895 | |||
XM_017024437.1 | XP_016879926.1 | 6 | 534 | |||
GenBank mRNA | AK123115.1 | - | 5 | 821 | ||
AK225602.1 | - | 6 | 478 | |||
AK308569.1 | - | 6 | 886 | |||
AK312958.1 | - | 6 | 478 | |||
AL050269.1 | - | 6 | 485 | |||
AL537925.3 | - | 6 | 685 | |||
AY632082.1 | - | 5 | 444 | |||
BC004195.2 | - | 6 | 514 | 3528894 | ||
BC004225.2 | - | 6 | 503 | 3538186 | ||
BI821280.1 | - | 7 | 509 | |||
BM909550.1 | - | 6 | 492 | |||
BP231776.1 | - | 6 | 526 | |||
BU159218.1 | - | 6 | 500 |