Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290261.1 | NP_001277190.1 | 4 | 1423 | ||
NM_015655.3 | NP_056470.1 | 5 | 1096 | |||
XM_005260702.3 | XP_005260759.1 | 4 | 912 | |||
XM_006723558.3 | XP_006723621.1 | 5 | 2099 | |||
XM_011529219.2 | XP_011527521.1 | 5 | 1219 | |||
XM_017027802.1 | XP_016883291.1 | 3 | 2994 | |||
XM_017027803.1 | XP_016883292.1 | 4 | 2003 | |||
XM_017027804.1 | XP_016883293.1 | 2 | 772 | |||
GenBank mRNA | AK056184.1 | - | 5 | 1007 | ||
AK056466.1 | - | 1 | 619 | |||
AK125455.1 | - | 4 | 1426 | |||
AK291235.1 | - | 5 | 1033 | |||
AK299811.1 | - | 3 | 1098 | |||
AL049942.1 | - | 1 | 137 | |||
BC021298.2 | - | 5 | 1025 | 4551314 |