Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015658.3 | NP_056473.2 | 9 | 1023 | ||
GenBank mRNA | AK022756.1 | - | 3 | 266 | ||
AK024284.1 | - | 9 | 977 | |||
AK225239.1 | - | 9 | 978 | |||
AK315080.1 | - | 9 | 992 | |||
AL050019.1 | - | 9 | 979 | |||
AW749585.1 | - | 1 | 70 | |||
BC003555.1 | - | 9 | 996 | 3546209 |
Gene Symbol: NOC2LP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002826.2 | 1 | 974 | |||
GenBank mRNA | AK302229.1 | - | 1 | 974 | ||
BC060851.1 | - | 1 | 967 |