Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSL1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015659.2 | NP_056474.2 | 5, 6 | 716 | ||
GenBank mRNA | AF083127.1 | - | 4, 5 | 419 | ||
AJ007398.1 | - | 5, 6 | 648 | |||
AK000822.1 | - | 5, 6 | 660 | |||
AK295935.1 | - | 4, 5 | 571 | |||
AL049999.1 | - | 5, 6 | 702 | |||
AY154473.1 | - | 5, 6 | 677 | |||
AY598331.1 | - | 5, 6 | 643 | |||
BC016362.1 | - | 5, 6 | 679 | |||
BC017899.1 | - | 5, 6 | 665 | |||
BC019069.2 | - | 5, 6 | 649 | |||
BC070325.1 | - | 5, 6 | 667 | |||
BC107782.1 | - | 5, 6 | 669 | |||
BC112228.1 | - | 5, 6 | 681 | 8327696 | ||
BC113699.1 | - | 5, 6 | 669 | 8322751 | ||
BI856275.1 | - | 5, 6 | 717 |