Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015662.2 | NP_056477.1 | 27, 28 | 3141 | ||
XM_005264254.1 | XP_005264311.1 | 27, 28 | 2972 | |||
XM_006711986.3 | XP_006712049.1 | 28, 29 | 3233 | |||
XM_006711987.1 | XP_006712050.1 | 27, 28 | 3038 | |||
XM_011532757.2 | XP_011531059.1 | 27, 28 | 3347 | |||
XM_011532758.1 | XP_011531060.1 | 27, 28 | 3038 | |||
XM_011532759.2 | XP_011531061.1 | 13, 14 | 1623 | |||
XM_011532760.2 | XP_011531062.1 | 10, 11 | 1303 | |||
XM_017003790.1 | XP_016859279.1 | 28, 29 | 3167 | |||
XM_017003791.1 | XP_016859280.1 | 26, 27 | 3108 | |||
XM_017003792.1 | XP_016859281.1 | 27, 28 | 3038 | |||
XM_017003793.1 | XP_016859282.1 | 23, 24 | 2548 | |||
XM_017003794.1 | XP_016859283.1 | 21, 22 | 2251 | |||
XM_017003795.1 | XP_016859284.1 | 25, 26 | 3100 | |||
GenBank mRNA | AB033005.1 | - | 9, 10 | 1006 | ||
AL110218.1 | - | 24, 25 | 2697 | |||
BC137126.1 | - | 27, 28 | 3031 | 9021123 |