Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015666.3 | NP_056481.1 | 7 | 1000 | ||
XM_005260393.1 | XP_005260450.1 | 7 | 990 | |||
XM_006723774.3 | XP_006723837.1 | 8 | 1084 | |||
XM_011528776.1 | XP_011527078.1 | 7 | 3004 | |||
XM_017027805.1 | XP_016883294.1 | 6 | 3154 | |||
XM_017027806.1 | XP_016883295.1 | 7 | 989 | |||
XM_017027807.1 | XP_016883296.1 | 6 | 3180 | |||
XM_017027808.1 | XP_016883297.1 | 7 | 1016 | |||
GenBank mRNA | AK001603.1 | - | 6 | 980 | ||
AK124572.1 | - | 7 | 3291 | |||
AK299147.1 | - | 5 | 658 | |||
AK299726.1 | - | 4 | 509 | |||
AK316027.1 | - | 6 | 780 | |||
AK316325.1 | - | 7 | 852 | |||
AL117498.1 | - | 1 | 29 | |||
BC012073.1 | - | 5 | 865 | |||
BC036716.2 | - | 6 | 974 | 4900170 |