Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136017.3 | NP_001129489.1 | 3, 4 | 771 | ||
NM_001136125.2 | NP_001129597.1 | 2, 3 | 558 | |||
NM_001136126.2 | NP_001129598.1 | 2, 3 | 555 | |||
NM_001287427.1 | NP_001274356.1 | 3, 4 | 1274 | |||
NM_001287434.1 | NP_001274363.1 | 3, 4 | 737 | |||
NM_001760.4 | NP_001751.1 | 3, 4 | 774 | |||
GenBank mRNA | AB209825.1 | - | 3, 4 | 565 | ||
AK055519.1 | - | 3, 4 | 832 | |||
AK057206.1 | - | 3, 4 | 565 | |||
AK096276.1 | - | 3, 4 | 1085 | |||
AK097856.1 | - | 2, 3 | 349 | |||
AK315421.1 | - | 3, 4 | 690 | |||
BC011616.2 | - | 3, 4 | 656 | 3946312 | ||
CR542246.1 | - | 3, 4 | 575 | |||
DA069566.1 | - | 2, 3 | 374 | |||
DB110132.1 | - | 3, 4 | 531 | |||
DC360231.1 | - | 2, 3 | 465 | |||
M90814.1 | - | 3, 4 | 675 | |||
M92287.1 | - | 3, 4 | 740 |