Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012384.4 | NP_036516.1 | 4 | 544 | ||
XM_005260202.4 | XP_005260259.1 | 4 | 755 | |||
XM_006723776.2 | XP_006723839.1 | 2 | 196 | |||
XM_011528779.2 | XP_011527081.1 | 2 | 260 | |||
GenBank mRNA | AB033095.1 | - | 4 | 421 | ||
AF173867.1 | - | 3 | 820 | |||
AL133646.1 | - | 2 | 268 | |||
BC036305.1 | - | 4 | 471 | 5166938 | ||
BQ954281.1 | - | 3 | 246 |