Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012384.4 | NP_036516.1 | 5 | 576 | ||
XM_005260202.4 | XP_005260259.1 | 5 | 787 | |||
XM_006723776.2 | XP_006723839.1 | 3 | 228 | |||
XM_011528779.2 | XP_011527081.1 | 3 | 292 | |||
GenBank mRNA | AB033095.1 | - | 5 | 453 | ||
AF173867.1 | - | 4 | 852 | |||
AL133646.1 | - | 3 | 300 | |||
BC036305.1 | - | 5 | 503 | 5166938 | ||
BQ954281.1 | - | 4 | 278 |