Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001238.3 | NP_001229.1 | 9 | 937 | ||
NM_001322261.1 | NP_001309190.1 | 8 | 790 | |||
NM_001322262.1 | NP_001309191.1 | 8 | 890 | |||
XM_011527440.2 | XP_011525742.1 | 9 | 823 | |||
GenBank mRNA | AK291549.1 | - | 9 | 937 | ||
BC035498.2 | - | 9 | 927 | 3841192 | ||
KF672848.1 | - | 8 | 790 | |||
M73812.1 | - | 7 | 766 | |||
M74093.1 | - | 7 | 734 |