Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004055.1 | NP_001004055.1 | 7 | 1273 | ||
NM_012163.2 | NP_036295.1 | 7 | 1438 | |||
XM_017023126.1 | XP_016878615.1 | 6 | 1796 | |||
XM_017023127.1 | XP_016878616.1 | 6 | 2047 | |||
XM_017023128.1 | XP_016878617.1 | 6 | 1852 | |||
XM_017023129.1 | XP_016878618.1 | 6 | 2938 | |||
XM_017023130.1 | XP_016878619.1 | 6 | 1520 | |||
XM_017023131.1 | XP_016878620.1 | 6 | 1379 | |||
XM_017023132.1 | XP_016878621.1 | 6 | 1682 | |||
XM_017023133.1 | XP_016878622.1 | 7 | 1614 | |||
XM_017023134.1 | XP_016878623.1 | 6 | 1487 | |||
GenBank mRNA | AF176701.1 | - | 6 | 1377 | ||
AK096101.1 | - | 5 | 1910 | |||
BC018785.1 | - | 7 | 1438 | 4875646 |