Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM50B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012135.2 | NP_036267.1 | 2 | 1571 | ||
XM_017010729.1 | XP_016866218.1 | 2 | 1554 | |||
GenBank mRNA | BC001261.1 | - | 2 | 1549 | 3457886 | |
DB566944.1 | - | 1 | 373 | |||
Y18503.1 | - | 2 | 1539 |