Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257194.1 | NP_001244123.1 | 7 | 1273 | ||
NM_001257195.1 | NP_001244124.1 | 5 | 849 | |||
NM_017415.2 | NP_059111.2 | 7 | 1098 | |||
GenBank mRNA | AB032955.1 | - | 7 | 868 | ||
AF208068.1 | - | 7 | 782 | |||
AF208069.1 | - | 5 | 886 | |||
AF208070.1 | - | 7 | 1273 | |||
AK300172.1 | - | 2 | 279 | |||
AK303916.1 | - | 5 | 844 | |||
AK310568.1 | - | 6 | 768 | |||
AK311594.1 | - | 2 | 244 | |||
AK314707.1 | - | 7 | 839 | |||
BC034035.1 | - | 7 | 1097 | 4817650 |