Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLOC1S6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001311255.1 | NP_001298184.1 | 3 | 391 | ||
NM_012388.3 | NP_036520.1 | 3 | 557 | |||
NR_132350.1 | 4 | 565 | ||||
NR_132353.1 | 2 | 415 | ||||
NR_132356.1 | 3 | 259 | ||||
NR_132357.1 | 3 | 259 | ||||
GenBank mRNA | AF080470.1 | - | 3 | 461 | ||
AK128626.1 | - | 4 | 534 | |||
AK315581.1 | - | 3 | 275 | |||
AL596952.1 | - | 3 | 330 | |||
BC004819.1 | - | 3 | 278 | 3450397 | ||
DA859617.1 | - | 3 | 207 | |||
DB212570.1 | - | 2 | 302 | |||
DB289156.1 | - | 3 | 202 |