Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLOC1S6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001311255.1 | NP_001298184.1 | 3 | 449 | ||
NM_012388.3 | NP_036520.1 | 3 | 615 | |||
NR_132350.1 | 4 | 623 | ||||
NR_132351.1 | 3 | 611 | ||||
NR_132353.1 | 2 | 473 | ||||
NR_132354.1 | 2 | 469 | ||||
NR_132356.1 | 3 | 317 | ||||
NR_132357.1 | 3 | 317 | ||||
GenBank mRNA | AF080470.1 | - | 3 | 519 | ||
AK057545.1 | - | 3 | 355 | |||
AK128626.1 | - | 4 | 592 | |||
AK315581.1 | - | 3 | 333 | |||
AL596952.1 | - | 3 | 388 | |||
BC004819.1 | - | 3 | 336 | 3450397 | ||
DA798595.1 | - | 2 | 241 | |||
DA859617.1 | - | 3 | 265 | |||
DB212570.1 | - | 2 | 360 | |||
DB289156.1 | - | 3 | 260 |