Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLOC1S6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001311255.1 | NP_001298184.1 | 5 | 570 | ||
NM_001311256.1 | NP_001298185.1 | 4 | 482 | |||
NM_012388.3 | NP_036520.1 | 5 | 736 | |||
NR_132350.1 | 6 | 744 | ||||
NR_132351.1 | 5 | 732 | ||||
NR_132352.1 | 4 | 648 | ||||
NR_132353.1 | 4 | 594 | ||||
NR_132354.1 | 4 | 590 | ||||
NR_132355.1 | 3 | 506 | ||||
NR_132356.1 | 5 | 438 | ||||
NR_132357.1 | 4 | 351 | ||||
NR_132358.1 | 3 | 340 | ||||
NR_132359.1 | 3 | 208 | ||||
GenBank mRNA | AF080470.1 | - | 5 | 640 | ||
AK057545.1 | - | 5 | 476 | |||
AK128626.1 | - | 6 | 713 | |||
AK315581.1 | - | 5 | 454 | |||
AL596952.1 | - | 5 | 509 | |||
BC004819.1 | - | 5 | 457 | 3450397 | ||
DA226593.1 | - | 3 | 278 | |||
DA230439.1 | - | 3 | 341 | |||
DA233328.1 | - | 3 | 197 | |||
DA237963.1 | - | 3 | 209 | |||
DA247079.1 | - | 4 | 368 | |||
DA523523.1 | - | 4 | 296 | |||
DA798595.1 | - | 4 | 362 | |||
DA859617.1 | - | 4 | 299 | |||
DB212570.1 | - | 4 | 481 | |||
DB287075.1 | - | 3 | 279 | |||
DB289156.1 | - | 5 | 381 |