Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006616.2 | NP_001006617.2 | 5 | 725 | ||
NM_012171.2 | NP_036303.1 | 5 | 725 | |||
NM_130465.4 | NP_569732.2 | 5 | 725 | |||
XM_005265876.3 | XP_005265933.1 | 3 | 507 | |||
GenBank mRNA | AK024427.1 | - | 4 | 2596 | ||
BC010405.2 | - | 5 | 547 | 3621871 | ||
BC067105.1 | - | 5 | 610 | 6646576 | ||
BI756001.1 | - | 3, 4 | 442 |