Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012347.4 | NP_036479.1 | 4 | 435 | ||
NM_033480.2 | NP_258441.1 | 5 | 685 | |||
NM_033481.3 | NP_258442.2 | 5 | 562 | |||
XM_005248995.4 | XP_005249052.1 | 5 | 369 | |||
XM_005248996.4 | XP_005249053.1 | 5 | 520 | |||
GenBank mRNA | AB209833.1 | - | 5 | 724 | ||
AF174597.1 | - | 1 | 24 | |||
AF176704.1 | - | 4 | 435 | |||
AK021559.1 | - | 5 | 721 | |||
AL137520.1 | - | 5 | 685 | |||
AY858798.1 | - | 5 | 727 | |||
BC000650.2 | - | 5 | 562 | 3345926 | ||
BC024973.2 | - | 2 | 106 |