Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012180.2 | NP_036312.2 | 2 | 978 | ||
XM_017008020.1 | XP_016863509.1 | 1 | 115 | |||
GenBank mRNA | AF115514.1 | - | 2 | 361 | ||
AF174596.1 | - | 2 | 194 | |||
AF201932.1 | - | 2 | 387 | |||
AF233224.1 | - | 2 | 233 | |||
AK026319.1 | - | 2 | 401 | |||
AK123740.1 | - | 2 | 991 | |||
AK314483.1 | - | 2 | 396 | |||
AL602916.1 | - | 2 | 393 | |||
AY358721.1 | - | 2 | 364 | |||
BC014679.2 | - | 2 | 380 | 3857760 | ||
BC014903.1 | - | 2 | 978 | |||
BC040456.1 | - | 2 | 394 | 4281341 | ||
KU178588.1 | - | 1 | 128 | |||
KU178589.1 | - | 1 | 128 |