Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012175.3 | NP_036307.2 | 4 | 443 | ||
NM_033406.2 | NP_208385.1 | 4 | 443 | |||
XM_011519980.1 | XP_011518282.1 | 4 | 449 | |||
XM_011519981.2 | XP_011518283.1 | 4 | 449 | |||
XM_011519982.1 | XP_011518284.1 | 5 | 503 | |||
XM_017017533.1 | XP_016873022.1 | 5 | 503 | |||
GenBank mRNA | AB209039.1 | - | 3 | 3828 | ||
AF176702.1 | - | 4 | 400 | |||
AK001943.1 | - | 4 | 434 | |||
AK128454.1 | - | 4 | 421 | |||
AK304866.1 | - | 4 | 438 | |||
AL136625.1 | - | 4 | 425 | |||
AL832826.1 | - | 3 | 1115 | |||
BC039291.1 | - | 4 | 443 | |||
BC046110.1 | - | 4 | 429 | 5549548 | ||
BG716664.1 | - | 4 | 446 |