Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TINF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099274.1 | NP_001092744.1 | 3 | 682 | ||
NM_012461.2 | NP_036593.2 | 3 | 682 | |||
XM_005267529.3 | XP_005267586.1 | 2 | 508 | |||
XM_011536642.2 | XP_011534944.1 | 3 | 654 | |||
GenBank mRNA | AF195512.1 | - | 3 | 602 | ||
AK023166.1 | - | 3 | 623 | |||
AK225379.1 | - | 3 | 626 | |||
AK294119.1 | - | 2 | 508 | |||
AK315084.1 | - | 3 | 570 | |||
BC005030.1 | - | 3 | 463 | 3690254 | ||
BC019343.1 | - | 3 | 640 | 3505009 | ||
BM549897.1 | - | 3 | 665 | |||
BX161478.1 | - | 3 | 615 |