Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271814.1 | NP_001258743.1 | 2 | 135 | ||
NM_012400.3 | NP_036532.1 | 2 | 135 | |||
GenBank mRNA | AF112982.1 | - | 2 | 95 | ||
AF188625.1 | - | 2 | 103 | |||
AK290406.1 | - | 2 | 96 | |||
AK301396.1 | - | 2 | 135 | |||
AK310156.1 | - | 2 | 105 | |||
AK311903.1 | - | 2 | 105 | |||
BC025706.1 | - | 2 | 84 | 5223912 | ||
DC295429.1 | - | 2 | 135 |