Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1RAPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017416.1 | NP_059112.1 | 9 | 1894 | ||
XM_011530905.2 | XP_011529207.1 | 7 | 902 | |||
XM_017029403.1 | XP_016884892.1 | 9 | 1312 | |||
GenBank mRNA | AF181285.1 | - | 7 | 1056 | ||
AF212016.1 | - | 8 | 1138 | |||
AF284436.1 | - | 9 | 1289 | |||
AJ272208.1 | - | 9 | 1894 | |||
AJ290436.1 | - | 8 | 1157 | |||
AK290196.1 | - | 9 | 2010 | |||
BC104784.1 | - | 9 | 1186 | 8143787 | ||
BC104786.1 | - | 9 | 1186 | 8143789 |