Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019851.2 | NP_062825.1 | 3 | 588 | ||
GenBank mRNA | AB030648.1 | - | 3 | 455 | ||
AB044277.1 | - | 3 | 588 | |||
BC096720.1 | - | 4 | 383 | |||
BC098128.1 | - | 4 | 384 | |||
BC098153.1 | - | 4 | 384 | |||
BC098339.1 | - | 4 | 384 | |||
BC127125.1 | - | 3 | 253 | |||
BC137446.1 | - | 3 | 526 | 9021446 | ||
BC137447.1 | - | 3 | 526 | 9021447 | ||
BC141979.1 | - | 3 | 283 | |||
BC141980.1 | - | 4 | 427 | |||
DT220794.1 | - | 3 | 385 |